Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyllithium, 2.2M (6wt%) in diethyl ether with LiBr, AcroSeal™, Thermo Scientific Chemicals

Catalog No. AC181291000
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC181291000 100 mL AcroSeal™ Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC181291000 Supplier Thermo Scientific Chemicals Supplier No. 181291000

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 917-54-4 , 60-29-7
Molecular Formula CH3Li
Molecular Weight (g/mol) 21.98
MDL Number MFCD00008253
InChI Key DVSDBMFJEQPWNO-UHFFFAOYSA-N
Synonym methyllithium, lithium, methyl, methyl lithium, lithium methanide, meli, lithium methyl, lithium carbanide, lithium methide, methllithium, methyllithum
PubChem CID 2724049
IUPAC Name lithium;carbanide
SMILES [Li]C

Specifications

Color Colorless through Yellow
Physical Form Solution
Quantity 100 mL
Linear Formula CH3Li
Flash Point −28°C
Solubility Information Solubility in water: vigorous reaction
Formula Weight 21.98
Percent Purity 9.0 to 11.0% (LiBr), 6.0 to 7.0% (LiCH3)
Packaging AcroSeal™ Glass bottle
Specific Gravity 0.81
Density 0.8100g/mL
Chemical Name or Material Methyllithium
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
May cause drowsiness or dizziness.
Causes severe skin burns and eye damage.
Catches fire spontaneously if exposed to air.
In contact with water releases flammable gases which may ignite
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p
EINECSNumber 213-026-4
Recommended Storage Product may develop some turbidity or precipitate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.