Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
(Methylthio)acetaldehyde Dimethyl Acetal 98.0+%, TCI America™

Click to view available options
Quantity:
5 g
25 g
Specifications
Specifications
Color | Yellow |
Boiling Point | 60°C |
Quantity | 25 g |
UN Number | 1993 |
Synonym | methylthio acetaldehyde dimethyl acetal, 1,1-dimethoxy-2-methylthio ethane, ethane, 1,1-dimethoxy-2-methylthio, 1,1-dimethoxy-2-methylsulfanyl-ethane, ethane,1,1-dimethoxy-2-methylthio, methylmercapto acetaldehyde dimethyl acetal, 1,1-dimethoxy-2-methylsulfanyl ethane, acmc-209j9v, methylthio acetaldehyde dimethylacetal, 2-methylthio acetaldehyde dimethyl acetal |
Formula Weight | 136.21 |
Percent Purity | ≥98.0% (GC) |
Physical Form | Liquid |
Chemical Name or Material | (Methylthio)acetaldehyde Dimethyl Acetal |
Safety and Handling
TSCA | No |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction