Learn More
Methyltrimethoxysilane, 97%
Description
Methyltrimethoxysilane is used as an acid scavenger, for example in the formation of substituted azulenes from allenylsilanes and tropylium tetrafluoroborate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Methyltrimethoxysilane is used as an acid scavenger, for example in the formation of substituted azulenes from allenylsilanes and tropylium tetrafluoroborate.
Solubility
Decomposes in water.
Notes
Moisture Sensitive. Store under dry inert gas. Store away from moisture, strong bases and oxidizing agents. Protect from humidity and water.
Specifications
Specifications
| Boiling Point | 102°C to 103°C |
| Quantity | 250 mL |
| UN Number | UN1993 |
| Beilstein | 1736151 |
| Flash Point | 9°C (48°F) |
| Solubility Information | Decomposes in water. |
| Formula Weight | 136.23 |
| Percent Purity | 97% |
| Refractive Index | 1.37 |
| Sensitivity | Moisture sensitive |
| Show More |
Safety and Handling
| Hazard Category | H225-H315-H319-H335 |
| Hazard Statement | GHS H Statement H225-H315-H319-H335 Highly flammable liquid and vapour. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S. |
| EINECSNumber | 214-685-0 |
| RTECSNumber | VV4650000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.