Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Methysergide maleate
GREENER_CHOICE

Catalog No. p-4783640
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10-641-0 10 mg
10-645-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 10-641-0 Supplier Tocris Bioscience™ Supplier No. 1064/10
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

5-HT1/5-HT2 antagonist

Mixed 5-HT1/5-HT2 receptor antagonist.

Chemical Identifiers

CAS 129-49-7
Molecular Formula C25H31N3O6

Specifications

Quantity 10 mg
Formula Weight 469.54
Purity >99%
Molecular Weight (g/mol) 469.54
Chemical Name or Material Methysergide maleate
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.