Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

METTL1 Antibody (9C2F2), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB350894 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB350894 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB350894 Supplier Novus Biologicals Supplier No. NBP306651MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

METTL1 Monoclonal antibody specifically detects METTL1 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation, ELISA

Specifications

Antigen METTL1
Applications Western Blot, ELISA, Immunoprecipitation, ELISA
Classification Monoclonal
Clone 9C2F2
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA, Immunoprecipitation, Sandwich ELISA Detection
Formulation 50mM Sodium Borate
Gene Alias C12orf1, D1075-like gene product, EC 2.1.1.33, FLJ95748, methyltransferase like 1, methyltransferase-like 1, Methyltransferase-like protein 1, TRM8, tRNA (guanine-N(7)-)-methyltransferase, tRNA(m7G46)-methyltransferase, YDL201w
Host Species Mouse
Immunogen This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human METTL1 (Accession#: NP_005362.3; Asp 32-Gln 265).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4234
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.