Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
METTL1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | METTL1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15492820
![]() |
Novus Biologicals
NBP15492820UL |
20 μL |
Each for $206.00
|
|
|||||
NBP154928
![]() |
Novus Biologicals
NBP154928 |
100 μL |
Each for $487.50
|
|
|||||
Description
METTL1 Polyclonal specifically detects METTL1 in Human samples. It is validated for Western Blot.Specifications
METTL1 | |
Polyclonal | |
Purified | |
RUO | |
4234 | |
Synthetic peptides corresponding to METTL1(methyltransferase like 1) The peptide sequence was selected from the middle region of METTL1. Peptide sequence KGQLTKMFFLFPDPHFKRTKHKWRIISPTLLAEYAYVLRVGGLVYTITDV. | |
Primary | |
34 kDa |
Western Blot | |
Unconjugated | |
Rabbit | |
C12orf1, D1075-like gene product, EC 2.1.1.33, FLJ95748, methyltransferase like 1, methyltransferase-like 1, Methyltransferase-like protein 1, TRM8, tRNA (guanine-N(7)-)-methyltransferase, tRNA(m7G46)-methyltransferase, YDL201w | |
METTL1 | |
IgG | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title