Learn More
Invitrogen™ METTL14 Monoclonal Antibody (CL4252)

Description
Immunogen sequence: RSWNMDSRLQE IRERQKLRRQ LLAQQLGAES ADSIGAVLNS KDEQREIAET RETCRASYDT SAPNAKRKYL DEGETDEDKM EEYKDELEMQ QDEE Highest antigen sequence identity to the following orthologs - mouse 92%, rat 92%.
Specifications
Specifications
| Antigen | METTL14 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | CL4252 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | METTL14 |
| Gene Accession No. | Q9HCE5 |
| Gene Alias | G430022H21Rik; hMETTL14; hypothetical protein zgc:77296; KIAA1627; methyltransferase like 14; methyltransferase-like protein 14; methyltransferase-like protein 14; N6-adenosine-methyltransferase subunit METTL14; Mettl14; mKIAA1627; N6-adenosine-methyltransferase non-catalytic subunit; N6-adenosine-methyltransferase subunit METTL14; RGD1304822; snoRNA MBII-84; zgc:77296 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.