Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

METTL14 Rabbit anti-Human, Alexa Fluor 594, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB049857 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB049857 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB049857 Supplier Novus Biologicals Supplier No. NBP297952AF594

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Polyclonal Antibody

METTL14 Polyclonal specifically detects METTL14 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen METTL14
Applications Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 594
Dilution Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias EC 2.1.1, EC 2.1.1.-, KIAA1627methyltransferase-like protein 14, methyltransferase like 14
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human METTL14 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 57721
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.