Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

METTL25 Antibody (OTI2B6), PE, Novus Biologicals™
SDP

Catalog No. NB287903 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB287903 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB287903 Supplier Novus Biologicals Supplier No. NBP272656PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

METTL25 Monoclonal antibody specifically detects METTL25 in Human samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen METTL25
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI2B6
Conjugate PE
Dilution Western Blot, Immunohistochemistry
Formulation PBS
Gene Alias C12orf26, chromosome 12 open reading frame 26, FLJ22789, hypothetical protein LOC84190
Host Species Mouse
Immunogen Full length human recombinant protein of human C12orf26(NP_115606) produced in HEK293T cell.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84190
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.