Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
METTL3 Monoclonal Antibody (29C8), Active Motif™

Mouse Monoclonal Antibody
$156.65
Specifications
Antigen | METTL3 |
---|---|
Clone | 29C8 |
Applications | Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-227-7272
![]() ![]() |
Active Motif
65688 |
10 μL |
Each of 1 for $156.65
|
|
|||||
Description
METTL3 Monoclonal antibody specifically detects METTL3 in Human samples. It is validated for Immunoprecipitation, Western BlotSpecifications
METTL3 | |
Immunoprecipitation, Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q86U44 | |
56339 | |
IgG2a | |
-20° C, Avoid Freeze/Thaw Cycles | |
METTL3 |
29C8 | |
Monoclonal | |
Liquid | |
RUO | |
tissue culture supernatant with 30% glycerol and 0.035% sodium azide | |
2310024F18Rik; adoMet-binding subunit of the human mRNA (N6-adenosine)-methyltransferase; fj82g07; hMETTL3; IME4; M6A; m6a methyltransferase; methyltransferase like 3; methyltransferase-like 3; methyltransferase-like protein 3; METTL3; mRNA (2'-O-methyladenosine-N(6)-)-methyltransferase; mRNA m(6)A methyltransferase; MTA70; MT-A70; N6-adenosine-methyltransferase 70 kDa subunit; N6-adenosine-methyltransferase catalytic subunit; N6-adenosine-methyltransferase subunit METTL3; Spo8; Unknown (protein for MGC:77093); wu:fj82g07; zgc:77093 | |
This antibody was raised against a peptide within the N-terminal region of human METTL3 (N6-adenosine-methyltransferase). | |
Primary | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title