Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEX3A Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB344175 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB344175 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB344175 Supplier Novus Biologicals Supplier No. NBP241092MFV450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MEX3A Polyclonal antibody specifically detects MEX3A in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen MEX3A
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias MEX-3A, MEX3A mex-3 homolog A, ring finger and KH domain containing 4, RING finger and KH domain-containing protein 4, RKHD4, RNA-binding protein MEX3A
Host Species Rabbit
Immunogen Antibody was raised against a 14 amino acid synthetic peptide from near the carboxy terminus of human Rkhd4. The immunogen is located within amino acids 420 - 470 of Rkhd4. Amino Acid Squence: AGLPRRPPGEPLQG
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 92312
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.