Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MFAP3L Antibody (1059143), PE, Novus Biologicals™
SDP

Catalog No. NB305277 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB305277 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB305277 Supplier Novus Biologicals Supplier No. FAB113251P
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MFAP3L Monoclonal specifically detects MFAP3L in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen MFAP3L
Applications Flow Cytometry
Classification Monoclonal
Clone 1059143
Conjugate PE
Dilution Flow Cytometry
Formulation PBS
Gene Alias HSD39, HSD-39, microfibrillar-associated protein 3-like, NYD-sp9, testis development protein NYD-SP9
Host Species Mouse
Immunogen Human embryonic kidney cell, HEK293-derived human MFAP3L, Met1-Met149, Accession # O75121
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9848
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.