Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MFAP4 Antibody (12D11), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NBP242638AI Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP242638AI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP242638AI Supplier Novus Biologicals Supplier No. NBP242638AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MFAP4 Monoclonal specifically detects MFAP4 in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen MFAP4
Applications Western Blot, ELISA
Classification Monoclonal
Clone 12D11
Conjugate Alexa Fluor 700
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias microfibril-associated glycoprotein 4, microfibrillar-associated protein 4
Gene Symbols MFAP4
Host Species Mouse
Immunogen Recombinant human MFAP4 (22-255aa) purified from E. coli
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4239
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.