Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MFF Rabbit anti-Human, Mouse, Bovine, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB151831 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB151831 100 μg
NB151832 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB151831 Supplier Novus Biologicals Supplier No. NBP311947100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MFF Polyclonal antibody specifically detects MFF in Human, Mouse, Bovine samples. It is validated for Western Blot, Immunofluorescence

Specifications

Antigen MFF
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:200
Formulation Neat Serum
Gene Alias mitochondrial fission factor
Host Species Rabbit
Immunogen Human recombinant protein. (UniProt: Q9GZY8)
Purification Method Unpurified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56947
Target Species Human, Mouse, Bovine
Content And Storage Store at -20C.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.