Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MFF Rabbit anti-Human, Mouse, Bovine, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
| Antigen | MFF |
|---|---|
| Dilution | Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:200 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
MFF Polyclonal antibody specifically detects MFF in Human, Mouse, Bovine samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
| MFF | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Bovine | |
| mitochondrial fission factor | |
| Human recombinant protein. (UniProt: Q9GZY8) | |
| Unpurified |
| Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Neat Serum | |
| 56947 | |
| Primary | |
| Store at -20C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title