Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MFG-E8 Antibody (278918), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB364250 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB364250 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB364250 Supplier Novus Biologicals Supplier No. FAB27671E
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MFG-E8 Monoclonal specifically detects MFG-E8 in Human samples. It is validated for CyTOF, Flow Cytometry (Intracellular Staining), Western Blot.

Specifications

Antigen MFG-E8
Applications CyTOF, Flow Cytometry (Intracellular Staining), Western Blot
Classification Monoclonal
Clone 278918
Conjugate DyLight 405
Dilution Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias BA46, Breast epithelial antigen BA46, EDIL1, hP47, HsT19888, lactadherin, medin, MFG1, MFG-E8, MFGM, Milk fat globule-EGF factor 8, milk fat globule-EGF factor 8 protein, O-acetyl disialoganglioside synthase, OAcGD3S, SED1, SPAG10, sperm associated antigen 10, sperm surface protein hP47
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human MFG-E8, Leu24-Cys387, Accession # Q08431
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 4240
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.