Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MFG-E8 Antibody (EDM45) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NBP247813A Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
0.2 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP247813A 0.1 mg
NBP247813B 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP247813A Supplier Novus Biologicals Supplier No. NBP2478130.1MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MFG-E8 Monoclonal specifically detects MFG-E8 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence, CyTOF-ready.

Specifications

Antigen MFG-E8
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence, CyTOF
Classification Monoclonal
Clone EDM45
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry : 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin : 0.5 - 1.0 ug/ml, Immunofluorescence : 1 - 2 ug/ml, CyTOF-ready
Formulation PBS with No Preservative
Gene Alias BA46, Breast epithelial antigen BA46, EDIL1, hP47, HsT19888, lactadherin, medin, MFG1, MFG-E8, MFGM, Milk fat globule-EGF factor 8, milk fat globule-EGF factor 8 protein, O-acetyl disialoganglioside synthase, OAcGD3S, SED1, SPAG10, sperm associated antigen 10, sperm surface protein hP47
Gene Symbols MFGE8
Host Species Mouse
Immunogen Delipidated human milk fat globule membrane preparation
Molecular Weight of Antigen 45 kDa
Purification Method Protein A or G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 4240
Test Specificity Recognizes a protein of 40-45kDa, identified as human milk fat globule membrane protein (HMFG). HMFG is present on normal human breast epithelial cells and cell lines derived from breast carcinomas, as well as to the outer surface of the human milk fat globule. HMFG is considered as a differentiation marker. It is useful as specific breast epithelial marker and can also provide a tool to study the role of the cell surface in normal and neoplastic mammary development.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.