Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MFSD11 Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB352380 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB352380 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB352380 Supplier Novus Biologicals Supplier No. NBP306336MFV450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MFSD11 Polyclonal antibody specifically detects MFSD11 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen MFSD11
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias ET, FLJ20226, FLJ22196, major facilitator superfamily domain containing 11, major facilitator superfamily domain-containing protein 11, protein ET, UNC93-like protein MFSD11
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human MFSD11 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79157
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.