Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MFSD2B Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB164374 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB164374 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB164374 Supplier Novus Biologicals Supplier No. NBP281737F
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MFSD2B Polyclonal antibody specifically detects MFSD2B in Human, Rat samples. It is validated for ELISA

Specifications

Antigen MFSD2B
Applications ELISA
Classification Polyclonal
Conjugate FITC
Formulation PBS
Gene Alias MFSD2B major facilitator superfamily domain containing 2B
Host Species Rabbit
Immunogen MFSD2B antibody was raised against a 15 amino acid synthetic peptide near the center of human MFSD2B. The immunogen is located within amino acids 240 - 290 of MFSD2B. Amino Acid Squence: VKERPDPSAPASGPG
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 388931
Target Species Human, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.