Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MG-H1 Antibody (1H7G5), Novus Biologicals™
SDP

Catalog No. NBP262810 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP262810 100 μg
NB019783 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP262810 Supplier Novus Biologicals Supplier No. NBP262810
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MG-H1 Monoclonal specifically detects MG-H1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoassay.

Specifications

Antigen MG-H1
Applications Immunocytochemistry, Immunofluorescence, In Situ Hybridization (ISH), Western Blot
Classification Monoclonal
Clone 1H7G5
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunoassay
Formulation PBS containing 0.1% BSA with 0.02% Sodium Azide
Gene Alias methylglyoxal 5-hydro-5-methylimidazolones
Host Species Mouse
Immunogen Nalpha-acetyl-Ndelta-(5-hydro-5-methyl)-4-imidazolone
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG2
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.