Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MGA Antibody, DyLight 405, Novus Biologicals™
SDP

Catalog No. NB052524 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB052524 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB052524 Supplier Novus Biologicals Supplier No. NBP298097V
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MGA Polyclonal specifically detects MGA in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen MGA
Applications Immunofluorescence
Classification Polyclonal
Conjugate DyLight 405
Dilution Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias FLJ12634, KIAA0518, MAD5, MAX dimerization protein 5, MAX gene associated, MAX gene-associated protein, MXD5
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human MGA fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23269
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.