Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MGAT2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$158.00 - $487.50
Specifications
| Antigen | MGAT2 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP1624420
![]() |
Novus Biologicals
NBP16244120UL |
20 μL |
Each for $158.00
|
|
|||||
NBP162441
![]() |
Novus Biologicals
NBP162441 |
100 μL |
Each for $487.50
|
|
|||||
Description
MGAT2 Polyclonal specifically detects MGAT2 in Human samples. It is validated for Western Blot.Specifications
| MGAT2 | |
| Polyclonal | |
| Rabbit | |
| Q10469 | |
| 4247 | |
| IgG | |
| 51 kDa |
| Western Blot | |
| Unconjugated | |
| RUO | |
| alpha-1,6-mannosyl-glycoprotein 2-beta-N-acetylglucosaminyltransferase, Beta-1,2-N-acetylglucosaminyltransferase II, CDG2A, EC 2.4.1.143, GlcNAc-T II, GLCNACTII, GNT2, GNT-IICDGS2, Mannoside acetylglucosaminyltransferase 2, mannosyl (alpha-1,6-)-glycoprotein beta-1,2-N-acetylglucosaminyltransferase, N-glycosyl-oligosaccharide-glycoprotein N-acetylglucosaminyltransferase II, UDP-N-acetylglucosamine:alpha-6-D-mannosidebeta-1,2-N-acetylglucosaminyltransferase II | |
| MGAT2 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title