Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MGC20410 Antibody (PCRP-BATF2-2B9), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP320601100UG
Description
MGC20410 Monoclonal antibody specifically detects MGC20410 in Human samples. It is validated for Flow Cytometry, ELISA, Immunoprecipitation, Immunohistochemistry (Paraffin)Specifications
MGC20410 | |
Monoclonal | |
0.2 mg/ml | |
Flow Cytometry 1-2 μg/million cells, ELISA, Immunoprecipitation 1-2 μg per 100-500 μg of protein or 1ml of lysate, Immunohistochemistry-Paraffin 1-2 μg/mL | |
basic leucine zipper transcription factor, ATF-like 2, basic leucine zipper transcriptional factor ATF-like 2, B-ATF-2, MGC20410, SARI, suppressor of AP-1 regulated by IFN | |
Recombinant fragment (around aa5-140) of human MGC20410 (exact sequence is proprietary) | |
100 μg | |
Primary | |
Human | |
Purified |
Flow Cytometry, ELISA, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
PCRP-BATF2-2B9 | |
Unconjugated | |
10mM PBS with 0.05% BSA | |
Mouse | |
Protein A or G purified | |
RUO | |
116071 | |
Store at 4&drg;C. Do not freeze. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction