Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MGC20410 Antibody (PCRP-BATF2-2B9), Novus Biologicals™

Mouse Monoclonal Antibody
$216.50 - $443.50
Specifications
Antigen | MGC20410 |
---|---|
Clone | PCRP-BATF2-2B9 |
Concentration | 0.2 mg/ml |
Dilution | Flow Cytometry 1-2 μg/million cells, ELISA, Immunoprecipitation 1-2 μg per 100-500 μg of protein or 1ml of lysate, Immunohistochemistry-Paraffin 1-2 μg/mL |
Applications | Flow Cytometry, ELISA, Immunoprecipitation, Immunohistochemistry (Paraffin) |
Description
MGC20410 Monoclonal antibody specifically detects MGC20410 in Human samples. It is validated for Flow Cytometry, ELISA, Immunoprecipitation, Immunohistochemistry (Paraffin)Specifications
MGC20410 | |
0.2 mg/ml | |
Flow Cytometry, ELISA, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Human | |
basic leucine zipper transcription factor, ATF-like 2, basic leucine zipper transcriptional factor ATF-like 2, B-ATF-2, MGC20410, SARI, suppressor of AP-1 regulated by IFN | |
Recombinant fragment (around aa5-140) of human MGC20410 (exact sequence is proprietary) | |
Primary | |
Store at 4&drg;C. Do not freeze. |
PCRP-BATF2-2B9 | |
Flow Cytometry 1-2 μg/million cells, ELISA, Immunoprecipitation 1-2 μg per 100-500 μg of protein or 1ml of lysate, Immunohistochemistry-Paraffin 1-2 μg/mL | |
Monoclonal | |
Purified | |
RUO | |
10mM PBS with 0.05% BSA | |
116071 | |
IgG2b κ | |
Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title