Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

mGluR1 Antibody (511601), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB347570 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB347570 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB347570 Supplier Novus Biologicals Supplier No. FAB48361MFV610
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

mGluR1 Monoclonal specifically detects mGluR1 in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen mGluR1
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 511601
Conjugate mFluor Violet 610 SE
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias glutamate receptor, metabotropic 1, GRM1A, metabotropic glutamate receptor 1, mGluR1, MGLUR1A, MGLUR1GPRC1AmGlu1
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human mGluR1a, Met1-Ser522, Accession # Q13255
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission, Vision
Primary or Secondary Primary
Gene ID (Entrez) 2911
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.