Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

mGluR1 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB10093555 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10093555 0.1 mg
NB017908 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10093555 Supplier Novus Biologicals Supplier No. NB10093555

Rabbit Polyclonal Antibody

mGluR1 Polyclonal specifically detects mGluR1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.

Specifications

Antigen metabotropic Glutamate Receptor 1a
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000, Immunohistochemistry 1:100-1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. Q13255
Gene Alias glutamate receptor, metabotropic 1, GRM1A, metabotropic glutamate receptor 1, mGluR1, MGLUR1A, MGLUR1GPRC1AmGlu1
Gene Symbols GRM1
Host Species Rabbit
Immunogen Synthetic peptide comprising internal residues of the human metabotropic GluR1 alpha protein. Reacts with rat and mouse GluR1.
Molecular Weight of Antigen 132.4 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline GPCR, Metabotropic Glutamate Receptors, Neuronal Cell Markers, Neuroscience, Neurotransmission, Vision
Primary or Secondary Primary
Gene ID (Entrez) 2911
Test Specificity mGluR1, Synthetic peptide comprising internal residues of the human GluR1 alpha protein. Reacts with rat and mouse mGluR1.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.