Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
mGluR1 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50
Specifications
Antigen | metabotropic Glutamate Receptor 1a |
---|---|
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500-1:1000, Immunohistochemistry 1:100-1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen |
Applications | Western Blot, Immunohistochemistry (Frozen), Immunocytochemistry, Immunohistochemistry, Immunofluorescence |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017908
![]() |
Novus Biologicals
NB10093555SS |
0.025 mg |
Each for $206.50
|
|
|||||
NB10093555
![]() |
Novus Biologicals
NB10093555 |
0.1 mg | N/A | N/A | N/A | ||||
Description
mGluR1 Polyclonal specifically detects mGluR1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.Specifications
metabotropic Glutamate Receptor 1a | |
Western Blot 1:500-1:1000, Immunohistochemistry 1:100-1:500, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen | |
Polyclonal | |
Rabbit | |
GPCR, Metabotropic Glutamate Receptors, Neuronal Cell Markers, Neuroscience, Neurotransmission, Vision | |
PBS with 0.02% Sodium Azide | |
glutamate receptor, metabotropic 1, GRM1A, metabotropic glutamate receptor 1, mGluR1, MGLUR1A, MGLUR1GPRC1AmGlu1 | |
GRM1 | |
IgG | |
Affinity Purified | |
mGluR1, Synthetic peptide comprising internal residues of the human GluR1 alpha protein. Reacts with rat and mouse mGluR1. |
1 mg/mL | |
Western Blot, Immunohistochemistry (Frozen), Immunocytochemistry, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
Human, Rat, Mouse | |
Q13255 | |
2911 | |
Synthetic peptide comprising internal residues of the human metabotropic GluR1 alpha protein. Reacts with rat and mouse GluR1. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
132.4 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title