Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

mGluR1 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB344904 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB344904 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB344904 Supplier Novus Biologicals Supplier No. NB11039033PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

mGluR1 Polyclonal antibody specifically detects mGluR1 in Human, Mouse, Rat, Chicken samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)

Specifications

Antigen mGluR1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation PBS
Gene Alias glutamate receptor, metabotropic 1, GRM1A, metabotropic glutamate receptor 1, mGluR1, MGLUR1A, MGLUR1GPRC1AmGlu1
Host Species Rabbit
Immunogen Synthetic peptide comprising residues 1159-1171 [SSVPSSPVSESVL] of the human mGluR1 protein.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission, Vision
Primary or Secondary Primary
Gene ID (Entrez) 2911
Target Species Human, Mouse, Rat, Chicken
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.