Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

mGluR2 Antibody (455310), Biotin, Novus Biologicals™
SDP

Catalog No. NB369355 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB369355 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB369355 Supplier Novus Biologicals Supplier No. FAB4676B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

mGluR2 Monoclonal specifically detects mGluR2 in Human samples. It is validated for CyTOF, Flow Cytometry (Intracellular Staining), Western Blot.

Specifications

Antigen mGluR2
Applications CyTOF, Flow Cytometry (Intracellular Staining), Western Blot
Classification Monoclonal
Clone 455310
Conjugate Biotin
Dilution Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias GLUR2, glutamate metabotropic receptor 2, glutamate receptor, metabotropic 2, GPRC1Bmetabotropic glutamate receptor 2, mGlu2, mGluR2, MGLUR2glutamate receptor homolog
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human mGluR2, Glu19-Ser498, Accession # Q14416
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission, Tumor Suppressors, Vision
Primary or Secondary Primary
Gene ID (Entrez) 2912
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.