Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

mGluR3 Antibody (530503), Biotin, Novus Biologicals™
SDP

Catalog No. NB369417 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB369417 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB369417 Supplier Novus Biologicals Supplier No. FAB4668B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

mGluR3 Monoclonal specifically detects mGluR3 in Human samples. It is validated for CyTOF, Flow Cytometry, Immunohistochemistry.

Specifications

Antigen mGluR3
Applications CyTOF, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone 530503
Conjugate Biotin
Dilution Flow Cytometry, Immunohistochemistry, CyTOF-ready
Formulation PBS
Gene Alias GLUR3, glutamate receptor, metabotropic 3, GPRC1Cglutamate metabotropic receptor 3, GRM3, mGlu3, mGluR3, MGLUR3metabotropic glutamate receptor 3
Host Species Mouse
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human mGluR3, Asp25-Ser507, Accession # Q14832
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission, Vision
Primary or Secondary Primary
Gene ID (Entrez) 2913
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.