Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
mGluR4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP303682100UL
Description
mGluR4 Polyclonal specifically detects mGluR4 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
mGluR4 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
glutamate receptor, metabotropic 4, GPRC1Dmetabotropic glutamate receptor 4, MGC177594, mGlu4, mGluR4, MGLUR4glutamate metabotropic receptor 4 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 33-290 of human mGluR4 (NP_000832.1). | |
100 μL | |
GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission, Vision | |
2914 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction