Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
mGluR4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $499.50
Specifications
Antigen | mGluR4 |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
mGluR4 Polyclonal specifically detects mGluR4 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
mGluR4 | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
glutamate receptor, metabotropic 4, GPRC1Dmetabotropic glutamate receptor 4, MGC177594, mGlu4, mGluR4, MGLUR4glutamate metabotropic receptor 4 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 33-290 of human mGluR4 (NP_000832.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Rabbit | |
GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission, Vision | |
PBS with 50% glycerol, pH7.3. | |
2914 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title