Learn More
Description
Specifications
Specifications
| Antigen | metabotropic Glutamate Receptor 5/1 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
| Classification | Monoclonal |
| Clone | S75-3 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1ug/ml, Immunohistochemistry 0.1-1.0ug/ml, Immunocytochemistry/Immunofluorescence 0.1-1.0ug/ml, Immunoprecipitation 1:10-1:500 |
| Gene Accession No. | P31424 |
| Gene Alias | glutamate receptor, metabotropic 5, GPRC1EMGLUR5metabotropic glutamate receptor 5, mGlu5, mGluR5 |
| Gene Symbols | GRM5 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
