Learn More
mGluR5/1a Antibody (S75-33), Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP259334
 View more versions of this product
Catalog No. NBP259334
Only null left
Add to Cart
Description
Description
mGluR5/1a Monoclonal specifically detects mGluR5/1a in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
Specifications
Specifications
mGluR5/1a
Monoclonal
Unconjugated
PBS pH7.4, 50% glycerol with 0.09% Sodium Azide
Fusion protein amino acids 824-1203 (cytoplasmic C-terminus) of rat mGluR5b
100 μg
Primary
Human, Mouse, Rat
Purified
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
S75-33
Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500
Mouse
Protein G purified
RUO
Detects 130kDa (both mGluR1 and mGluR5).
Store at -20C.
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only