Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
mGluR5/1a Antibody (S75-33), Novus Biologicals™

Mouse Monoclonal Antibody
$560.50
Specifications
Antigen | mGluR5/1a |
---|---|
Clone | S75-33 |
Dilution | Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500 |
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
Classification | Monoclonal |
Description
mGluR5/1a Monoclonal specifically detects mGluR5/1a in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
mGluR5/1a | |
Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500 | |
Monoclonal | |
Purified | |
RUO | |
Fusion protein amino acids 824-1203 (cytoplasmic C-terminus) of rat mGluR5b | |
Primary | |
Detects 130kDa (both mGluR1 and mGluR5). |
S75-33 | |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat | |
IgG1 | |
Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title