Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

mGluR6 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB338869 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB338869 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB338869 Supplier Novus Biologicals Supplier No. NB300189PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

mGluR6 Polyclonal antibody specifically detects mGluR6 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Frozen)

Specifications

Antigen mGluR6
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Frozen
Formulation PBS
Gene Alias CSNB1B, glutamate receptor, metabotropic 6, GPRC1Fmetabotropic glutamate receptor 6, mGlu6, mGluR6, MGLUR6DKFZp686H1993
Host Species Rabbit
Immunogen C-terminus of rat metabotropic Glutamate Receptor 6 (mGluR6) [Uniprot: P35349]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Amino Acids Drugs and other small molecules, GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission, Sensory Systems, Signal Transduction, Vision
Primary or Secondary Primary
Gene ID (Entrez) 2916
Target Species Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.