Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

mGluR8 Antibody (4C1C1) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP261857 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP2-61857 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP2-61857 Supplier Novus Biologicals Supplier No. NBP261857
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

mGluR8 Monoclonal specifically detects mGluR8 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.

Specifications

Antigen mGluR8
Applications Western Blot, Flow Cytometry, ELISA
Classification Monoclonal
Clone 4C1C1
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000
Formulation PBS with 0.05% Sodium Azide
Gene Alias FLJ41058, glutamate metabotropic receptor 8, glutamate receptor, metabotropic 8, GPRC1HMGLUR8GLUR8, metabotropic glutamate receptor 8, MGC126724, mGlu8, mGluR8
Gene Symbols GRM8
Host Species Mouse
Immunogen Purified recombinant fragment of human GRM8 (AA: extra 440-583) expressed in E. Coli.
Molecular Weight of Antigen 101.7 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline GPCR, Metabotropic Glutamate Receptors, Neuronal Cell Markers, Neuroscience, Neurotransmission
Primary or Secondary Primary
Gene ID (Entrez) 2918
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.