Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MGMT Antibody (MT 23.2), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB100168IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100168IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100168IR Supplier Novus Biologicals Supplier No. NB100168IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MGMT Monoclonal specifically detects MGMT in Human, Mouse (Negative) samples. It is validated for CyTOF, Flow Cytometry, Flow Cytometry (Intracellular Staining), Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen MGMT
Applications CyTOF, Flow Cytometry, Flow Cytometry (Intracellular Staining), Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone MT 23.2
Conjugate DyLight 755
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias 6-O-methylguanine-DNA methyltransferase, EC 2.1.1.63, methylated-DNA--protein-cysteine methyltransferase, methylguanine-DNA methyltransferase, O-6-methylguanine-DNA methyltransferase, O6-methylguanine-DNA methyltransferase, O-6-methylguanine-DNA-alkyltransferase
Gene Symbols MGMT
Host Species Mouse
Immunogen Recombinant human MGMT (O6-methylguanine-DNA methyltransferase) purified from E. coli [UniProt# P16455]
Molecular Weight of Antigen 24 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Chromatin Research, Direct Reversal of DNA Damage, DNA Repair, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 4255
Target Species Human, Mouse (Negative)
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.