Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MGMT Antibody (MT 3.1), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 6 publications
Supplier: Novus Biologicals NB100692
Description
MGMT Monoclonal specifically detects MGMT in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
MGMT | |
Monoclonal | |
Unconjugated | |
P16455 | |
MGMT | |
Recombinant human MGMT protein. | |
0.5 mL | |
Apoptosis, Cancer, Chromatin Research, Direct Reversal of DNA Damage, DNA Repair, Tumor Suppressors | |
4255 | |
Human | |
Supernatant |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
MT 3.1 | |
Western Blot 1:100-1:2000, Immunohistochemistry 1:25-1:50, Immunohistochemistry-Paraffin 1:25-1:50 | |
6-O-methylguanine-DNA methyltransferase, EC 2.1.1.63, methylated-DNA--protein-cysteine methyltransferase, methylguanine-DNA methyltransferase, O-6-methylguanine-DNA methyltransferase, O6-methylguanine-DNA methyltransferase, O-6-methylguanine-DNA-alkyltransferase | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
This stains all mantle zone lymphocytes and 50% of germinal center lymphocytes. Basaloid epithelial cells of tonsil squamous mucosa were also stained positive with this. | |
Store at 4°C. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction