Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MGMT Antibody (MT 3.1), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 6 publications
$268.00 - $878.00
Specifications
Antigen | MGMT |
---|---|
Clone | MT 3.1 |
Applications | Immunohistochemistry (Paraffin), Immunohistochemistry, Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017791
![]() |
Novus Biologicals
NB1006920.1ML |
0.1 mL |
Each for $268.00
|
|
|||||
NB100692025
![]() |
Novus Biologicals
NB1006920.25ML |
0.25 mL |
Each for $499.50
|
|
|||||
NB100692
![]() |
Novus Biologicals
NB100692 |
0.5 mL |
Each for $878.00
|
|
|||||
Description
MGMT Monoclonal specifically detects MGMT in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
MGMT | |
Immunohistochemistry (Paraffin), Immunohistochemistry, Western Blot | |
Unconjugated | |
Human | |
6-O-methylguanine-DNA methyltransferase, EC 2.1.1.63, methylated-DNA--protein-cysteine methyltransferase, methylguanine-DNA methyltransferase, O-6-methylguanine-DNA methyltransferase, O6-methylguanine-DNA methyltransferase, O-6-methylguanine-DNA-alkyltransferase | |
MGMT | |
IgG1 | |
This stains all mantle zone lymphocytes and 50% of germinal center lymphocytes. Basaloid epithelial cells of tonsil squamous mucosa were also stained positive with this. |
MT 3.1 | |
Monoclonal | |
Mouse | |
P16455 | |
4255 | |
Recombinant human MGMT protein. | |
Primary |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title