Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MGP Antibody (OTI11G6), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NP272660A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP272660A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP272660A70 Supplier Novus Biologicals Supplier No. NBP272660AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MGP Monoclonal specifically detects MGP in Human samples. It is validated for Western Blot.

Specifications

Antigen MGP
Applications Western Blot
Classification Monoclonal
Clone OTI11G6
Conjugate Alexa Fluor 750
Dilution Western Blot
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias Cell growth-inhibiting gene 36 protein, matrix Gla protein, MGLAP, NTI
Gene Symbols MGP
Host Species Mouse
Immunogen Full length recombinant protein of human MGP (NP_000891) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 4256.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.