Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MGRN1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP149929 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP149929 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP149929 Supplier Novus Biologicals Supplier No. NBP149929
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MGRN1 Polyclonal specifically detects MGRN1 in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen MGRN1
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 ug/mg lysate
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Alias E3 ubiquitin-protein ligase MGRN1, EC 6.3.2, KIAA0544probable E3 ubiquitin-protein ligase MGRN1, Mahogunin RING finger protein 1, mahogunin, ring finger 1, RING finger protein 156, RNF156EC 6.3.2.-
Gene Symbols MGRN1
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 400 and 450 of human Mahogunin, Ring Finger 1 using the numbering given in entry NP_001135761.1 (GeneID 23295).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 23295
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.