Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MGRN1 Antibody (OTI3C3), PerCP, Novus Biologicals™
SDP

Catalog No. NB272665PCP Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB272665PCP 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB272665PCP Supplier Novus Biologicals Supplier No. NBP272665PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MGRN1 Monoclonal specifically detects MGRN1 in Human, Mouse, Rat, Monkey, Primate samples. It is validated for Flow Cytometry.

Specifications

Antigen MGRN1
Applications Flow Cytometry
Classification Monoclonal
Clone OTI3C3
Conjugate PerCP
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias E3 ubiquitin-protein ligase MGRN1, EC 6.3.2, KIAA0544probable E3 ubiquitin-protein ligase MGRN1, Mahogunin RING finger protein 1, mahogunin, ring finger 1, RING finger protein 156, RNF156EC 6.3.2.-
Gene Symbols MGRN1
Host Species Mouse
Immunogen Full length recombinant protein of human MGRN1 (NP_056061) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 23295.0
Target Species Human, Mouse, Rat, Monkey, Primate
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.