Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MHC Class I Antibody (2G5), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB10065938U Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10065938U 0.125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10065938U Supplier Novus Biologicals Supplier No. NB10065938UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MHC Class I Monoclonal specifically detects MHC Class I in Mouse samples. It is validated for Flow Cytometry, Immunoprecipitation.

Specifications

Antigen MHC Class I
Applications Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone 2G5
Conjugate DyLight 350
Dilution Flow Cytometry
Gene Alias DKFZp686P19218, EA1.2, EA2.1, HLA class I histocompatibility antigen, alpha chain E, HLA class I histocompatibility antigen, E alpha chain, HLA-6.2, HLAE, lymphocyte antigen, major histocompatibility complex, class I, E, MHC, MHC class I antigen E, MHC HLA-E alpha-1, MHC HLA-E alpha-2.1, QA1
Gene Symbols HLA-E
Host Species Mouse
Immunogen Purified H-2Kb and H-2Db MHC-I molecules.
Purification Method Protein G purified
Quantity 0.125 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology, Major Histocompatibility Complexes
Primary or Secondary Primary
Gene ID (Entrez) 3133
Test Specificity NB100-65938 recognizes a monomorphic epitope on the mouse MHC class I antigen. The epitope recognized by clone 2G5 is conformation dependent and is reported to be phylogenetically conserved. Reactivity has been observed with some canine samples suggesting that this antibody may recognize a polymorphic epitope of canine MHC class 1.
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.