Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MHC Class I Antibody (2G5), Janelia Fluor™ 549, Novus Biologicals™
SDP

Catalog No. NB211970 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB211970 0.125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB211970 Supplier Novus Biologicals Supplier No. NB10065938JF549
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MHC Class I Monoclonal antibody specifically detects MHC Class I in Human, Mouse, Rat, Porcine, Bovine, Chinese Hamster, Guinea Pig, Hamster, Sheep samples. It is validated for Flow Cytometry

Specifications

Antigen MHC Class I
Applications Flow Cytometry
Classification Monoclonal
Clone 2G5
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias DKFZp686P19218, EA1.2, EA2.1, HLA class I histocompatibility antigen, alpha chain E, HLA class I histocompatibility antigen, E alpha chain, HLA-6.2, HLAE, lymphocyte antigen, major histocompatibility complex, class I, E, MHC, MHC class I antigen E, MHC HLA-E alpha-1, MHC HLA-E alpha-2.1, QA1
Host Species Mouse
Immunogen Purified H-2Kb and H-2Db MHC-I molecules.
Purification Method Protein G purified
Quantity 0.125 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Asthma, Cell Biology, Core ESC Like Genes, Diabetes Research, DNA Repair, DNA replication Transcription Translation and Splicing, Immunology, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3133
Target Species Human, Mouse, Rat, Pig, Bovine, Chinese Hamster, Guinea Pig, Hamster, Sheep
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.