Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MHC Class I Antibody (ER-HR52), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB164952AF Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB164952AF 0.125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB164952AF Supplier Novus Biologicals Supplier No. NB10064952AF405
Add to Cart
Add to Cart

Rat Monoclonal Antibody

MHC Class I Monoclonal specifically detects MHC Class I in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry.

Specifications

Antigen MHC Class I
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry
Classification Monoclonal
Clone ER-HR52
Conjugate Alexa Fluor 405
Dilution Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias DKFZp686P19218, EA1.2, EA2.1, HLA class I histocompatibility antigen, alpha chain E, HLA class I histocompatibility antigen, E alpha chain, HLA-6.2, HLAE, lymphocyte antigen, major histocompatibility complex, class I, E, MHC, MHC class I antigen E, MHC HLA-E alpha-1, MHC HLA-E alpha-2.1, QA1
Gene Symbols HLA-E
Host Species Rat
Immunogen Adherent FI (CBAXBL) bone marrow stromal cells
Purification Method Protein G purified
Quantity 0.125 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Asthma, Cell Biology, Diabetes Research, Immunology, Major Histocompatibility Complexes
Primary or Secondary Primary
Gene ID (Entrez) 3133
Test Specificity NB100-64952 recognizes a polymorphic determinant on the murine MHC (major histocompatibility complex) class I molecule. In the mouse, the MHC class I molecule is expressed on the cell surface of all nucleated cells but expression levels may differ according to cell type. Clone ER-MP52 specifically recognizes mouse strains with the haplotypes H-2b,w16 and H-2d,p,q. Mouse strains with the haplotypes H-2f,r,s,w17,w23,w27 show weak reactivity with this antibody.
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.