Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MHC Class I Antibody (MEM-E/08), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB500305646 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500305646 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB500305646 Supplier Novus Biologicals Supplier No. NB500305JF646
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MHC Class I Monoclonal specifically detects MHC Class I in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen MHCClassI
Applications Flow Cytometry
Classification Monoclonal
Clone MEM-E/08
Conjugate Janelia Fluor 646
Dilution Flow Cytometry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias DKFZp686P19218, EA1.2, EA2.1, HLA class I histocompatibility antigen, alpha chain E, HLA class I histocompatibility antigen, E alpha chain, HLA-6.2, HLAE, lymphocyte antigen, major histocompatibility complex, class I, E, MHC, MHC class I antigen E, MHC HLA-E alpha-1, MHC HLA-E alpha-2.1, QA1
Gene Symbols HLA-E
Host Species Mouse
Immunogen Bacterially expressed recombinant HLA-E refolded with beta2-microglobulin and peptide.
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Adaptive Immunity, Asthma, Cell Biology, Core ESC Like Genes, Diabetes Research, DNA Repair, DNA replication Transcription Translation and Splicing, Immunology, Major Histocompatibility Complexes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3133
Test Specificity This antibody (clone MEM-E/08) recognized native surface-expressed HLA-E, but not denaturated heavy chain of HLA-E. HLA-E belongs to the MHC Class I molecules (MHC Class Ib; nonclassical) and it is expressed on many types of the human cells. The results revealed that antibody is remarkably specific for HLA-E, only with weak cross-reactivity with following classical MHC Class I molecules (MHC Class Ia): HLA-A24, -B7, -B27, -B51, -B54, -C7.
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.