Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MHC Class I Antibody (OX18), PerCP, Novus Biologicals™
SDP

Catalog No. NB1206405P Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1206405P 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1206405P Supplier Novus Biologicals Supplier No. NB1206405PCP
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MHC Class I Monoclonal specifically detects MHC Class I in Rat samples. It is validated for Flow Cytometry, Flow (Intracellular).

Specifications

Antigen MHC Class I
Applications Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone OX18
Conjugate PerCP
Dilution Flow Cytometry, Flow (Intracellular)
Gene Alias DKFZp686P19218, EA1.2, EA2.1, HLA class I histocompatibility antigen, alpha chain E, HLA class I histocompatibility antigen, E alpha chain, HLA-6.2, HLAE, lymphocyte antigen, major histocompatibility complex, class I, E, MHC, MHC class I antigen E, MHC HLA-E alpha-1, MHC HLA-E alpha-2.1, QA1
Gene Symbols HLA-E
Host Species Mouse
Immunogen Rat spleen glycoproteins
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology, Major Histocompatibility Complexes
Primary or Secondary Primary
Gene ID (Entrez) 3133
Test Specificity Recognizes a monomorphic determinant of rat MHC Class I (RT1A), expressed by all rat strains. However, quantitative measurements suggest that not all of the class I molecules are recognised.
Target Species Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.