Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MHC Class I H-2Kk Antibody (16-3-22S), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NBP128202AF Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP128202AF 0.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP128202AF Supplier Novus Biologicals Supplier No. NBP128202AF405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MHC Class I H-2Kk Monoclonal specifically detects MHC Class I H-2Kk in Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen H-2Kk
Applications Flow Cytometry
Classification Monoclonal
Clone 16-3-22S
Conjugate Alexa Fluor 405
Dilution Flow Cytometry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias adrenomedullin binding protein, age-related maculopathy susceptibility 1, AHUS1, AMBP1, ARMD4, beta-1H, beta-1-H-globulin, CFHL3, complement factor H, factor H, factor H-like 1, FH, FHL1, H factor 1, H factor 1 (complement), H factor 2 (complement), HF, HF1ARMS1, HF2, HUSMGC88246
Gene Symbols H2-K1
Host Species Mouse
Immunogen C3H mouse splenocytes
Purification Method Protein A or G purified
Quantity 0.25 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14972
Test Specificity An epitope on mouse H-2Kk MHC class I alloantigen
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.