Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MHC Class I RT1Ac Antibody (OX-27), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB164951AF Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB164951AF 0.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB164951AF Supplier Novus Biologicals Supplier No. NB10064951AF405

Mouse Monoclonal Antibody

MHC Class I RT1Ac Monoclonal specifically detects MHC Class I RT1Ac in Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.

Specifications

Antigen MHC Class I RT1Ac
Applications Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone OX-27
Conjugate Alexa Fluor 405
Dilution Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias HLA-E
Gene Symbols HLA-E
Host Species Mouse
Immunogen PHA activated rat lymphocytes
Purification Method Protein G purified
Quantity 0.25 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity NB100-64951 recognises a polymorphic determinant (c haplotype) of rat Class I MHC Antigen (RT-1A).
Target Species Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.